Learn More
Meso Scale Discovery U-PLEX Interferon Combo, 5 plates, 96-well, Human immunoassay kit, 4-plex, Cytokines & Chemokines detection
SDP
Catalog No. NC2748498
Encompass
The MESO SCALE DISCOVERY U-PLEX Interferon Combo is an advanced immunoassay kit for the sensitive measurement of multiple human interferon biomarkers from a single sample. This kit is designed expressly for research use, providing a practical solution for cytokine and chemokine analysis in immunological and inflammatory research. Employing ECL detection technology, it ensures exceptional sensitivity and a wide detection range for analytes in the low picogram/mL scale. It enables the simultaneous measurement of key interferons in various sample types including serum and plasma.
  • Immunoassay kit for cytokine and chemokine measurement
  • Utilizes Electrochemiluminescence (ECL) detection for high sensitivity
  • Suitable for human samples, including serum and plasma
  • Can detect four analytes in a single test
  • Pack size includes 5 plates
  • 96-well plate format for ease of use
  • For research use only
Catalog No. NC2748498 Supplier Meso Scale Discovery Supplier No. K15094K2
The MESO SCALE DISCOVERY U-PLEX Interferon Combo is an advanced immunoassay kit for the sensitive measurement of multiple human interferon biomarkers from a single sample. This kit is designed expressly for research use, providing a practical solution for cytokine and chemokine analysis in immunological and inflammatory research. Employing ECL detection technology, it ensures exceptional sensitivity and a wide detection range for analytes in the low picogram/mL scale. It enables the simultaneous measurement of key interferons in various sample types including serum and plasma.
  • Immunoassay kit for cytokine and chemokine measurement
  • Utilizes Electrochemiluminescence (ECL) detection for high sensitivity
  • Suitable for human samples, including serum and plasma
  • Can detect four analytes in a single test
  • Pack size includes 5 plates
  • 96-well plate format for ease of use
  • For research use only