Learn More
EVIDENT SCIENTIFIC MIS INC U-POT Polarizer
Catalog No. NC1829300
Encompass
U-POT Polarizer for transmitted light, 45mm, fits in light well.
Catalog No. NC1829300 Supplier EVIDENT SCIENTIFIC MIS INC Supplier No. 3812600
U-POT Polarizer for transmitted light, 45mm, fits in light well.