Learn More
Bioworld Newborn Calf Serum (<14 days of age), 500 mL
Catalog No. 50254179
Encompass_Preferred
Newborn calf serum will support a wide variety of established cell lines and primary cultures which might otherwise require fetal bovine serum. Empirical testing is the most reliable method for determining usefulness for a particular line.SpecificationsSterile-filteredMycoplasma-tested and virus-screened Ships on dry ice. U.S. or N.Z. Origin (dependant on availiability)
Catalog No. 50-254-179 Supplier Bioworld Supplier No. 306110762
Newborn calf serum will support a wide variety of established cell lines and primary cultures which might otherwise require fetal bovine serum. Empirical testing is the most reliable method for determining usefulness for a particular line.SpecificationsSterile-filteredMycoplasma-tested and virus-screened Ships on dry ice. U.S. or N.Z. Origin (dependant on availiability)