Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Faceshields

Catalog No. 19825543 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Double matrix faceshield headgear (483000)
Matrix faceshield headgear
Matrix headgear
Lens Material:
Acetate
Polycarbonate
Polyester
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Lens Material
19-825-543 Double matrix faceshield headgear (483000) Polyester
17-242 Matrix faceshield headgear Acetate
17-244A Double matrix faceshield headgear (483000) Polycarbonate
17-244 Double matrix faceshield headgear (483000) Acetate
17-242A Matrix headgear Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-825-543 Supplier MCR Safety Supplier No. 494800
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers superior protection from liquid splashes and flying debris that would otherwise be damaging to the face. Includes plastic face shield. (Headgear must be purchased separately.)

Available in an assortment of brackets and faceshield sizes and materials.

Compliance

Meets ANSI Z87.1+ standards.

Specifications

Shape Flat
Certifications/Compliance ANSI Z87.1+, CSA Z94.1
Lens Tint Clear
For Use With (Equipment) Double matrix faceshield headgear (483000)
Lens Material Polyester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.