Learn More
Ibidi USA U-SLIDE 18 Well -Box: 15 (individually packed)
SDP
Catalog No. NC2205965
Encompass
A chambered coverslip with 18 wells for cell culture immunofluorescence and high-end microscopy. An all-in-one 18 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Ideal for high-resolution imaging through the # 1.5 polymer coverslip bottom with the highest optical quality. Suitable for various micrscopy techniques. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2205965 Supplier Ibidi USA Supplier No. 81816
A chambered coverslip with 18 wells for cell culture immunofluorescence and high-end microscopy. An all-in-one 18 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Ideal for high-resolution imaging through the # 1.5 polymer coverslip bottom with the highest optical quality. Suitable for various micrscopy techniques. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)