Learn More
Ibidi USA µ-Slide 18 Well Flat, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305825
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide 18 Well Flat is a convenient multi-well plate for quick and easy tests to probe experimental parameters like antibody dilution, seeding density or most effective drug concentrations. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-825 Supplier Ibidi USA Supplier No. 81826
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide 18 Well Flat is a convenient multi-well plate for quick and easy tests to probe experimental parameters like antibody dilution, seeding density or most effective drug concentrations. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box