Learn More
Ibidi USA µ-Slide 18 Well, Glass Bottom - #1.5H Glass Bottom Coverslip
SDP
Catalog No. NC1752257
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the µ-Slides and µ-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The µ-Slide 18 Well Glass Bottom is a convenient chambered coverslip with 18 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1752257 Supplier Ibidi USA Supplier No. 81817
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the µ-Slides and µ-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The µ-Slide 18 Well Glass Bottom is a convenient chambered coverslip with 18 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box