Learn More
Ibidi USA µ-Slide 4 Well Ph+, Glass Bottom - #1.5H Glass Bottom Coverslip
SDP
Catalog No. NC1451847
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the µ-Slides and µ-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The µ-Slide 4 Well Ph+ Glass Bottom is a convenient chambered coverslip with 4 wells for cell culture, immunofluorescence, and high-end microscopy. The µ-Slide 4 Well Ph+ improves the optical quality of phase contrast microscopy by suspending a plate in the center of the wells. When correctly filled, the plate eliminates the refraction of light due to meniscus formation compared to an open well. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1451847 Supplier Ibidi USA Supplier No. 80447
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the µ-Slides and µ-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The µ-Slide 4 Well Ph+ Glass Bottom is a convenient chambered coverslip with 4 wells for cell culture, immunofluorescence, and high-end microscopy. The µ-Slide 4 Well Ph+ improves the optical quality of phase contrast microscopy by suspending a plate in the center of the wells. When correctly filled, the plate eliminates the refraction of light due to meniscus formation compared to an open well. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box