Learn More
Ibidi USA U-SLIDE 8 wells, HIGH GRID 500 um, Box of 15
SDP
Catalog No. NC2079775
Encompass
A chambered coverslip with 8 individual wells, high walls, and an imprinted 500 µm cell location grid for use in immunofluorescence and high-end microscopy. Ideal for locating cells or cell clusters. Observation of the sample through the polymer coverslip using high resolution microscopy. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)Grid: Grid-500
This item is not returnable. View return policy
Catalog No. NC2079775 Supplier Ibidi USA Supplier No. 80806G500
A chambered coverslip with 8 individual wells, high walls, and an imprinted 500 µm cell location grid for use in immunofluorescence and high-end microscopy. Ideal for locating cells or cell clusters. Observation of the sample through the polymer coverslip using high resolution microscopy. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)Grid: Grid-500