Learn More
Ibidi USA U-SLIDE 8WL COLLAGEN I ST 1.5, (15 pieces per box)
SDP
Catalog No. NC2224409
Encompass
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Slide 8 Well is a convenient chambered coverslip with 8 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5 polymer coverslip, surface modficiation: Collagen I, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC2224409 Supplier Ibidi USA Supplier No. 80829
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Slide 8 Well is a convenient chambered coverslip with 8 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5 polymer coverslip, surface modficiation: Collagen I, sterilized, 15 pcs./box