Learn More
Ibidi USA µ-Slide 15 Well 3D Glass Bottom
SDP
Catalog No. NC2395456
Encompass
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Slide Angiogenesis is a cell culture product for tube formation, angiogenesis assays and direct cell culture. Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Glass Coverslip. #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilized
Catalog No. NC2395456 Supplier Ibidi USA Supplier No. 81507
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Slide Angiogenesis is a cell culture product for tube formation, angiogenesis assays and direct cell culture. Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Glass Coverslip. #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilized