Learn More
Ibidi USA µ-Slide Angiogenesis, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. 50305820
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide Angiogenesis is a cell culture product for tube formation, angiogenesis assays and direct cell culture. Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Polymer Coverslip. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-820 Supplier Ibidi USA Supplier No. 81501
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide Angiogenesis is a cell culture product for tube formation, angiogenesis assays and direct cell culture. Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Polymer Coverslip. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box