Learn More
Ibidi USA µ-Slide Chemotaxis, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305776
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide Chemotaxis is a tool for investigation of chemotaxis and migration of nonadherent cells in 3D gel matrices and adherent cells on 2D surfaces. The chamber’s geometry is optimized for analyzing chemotaxis by video microscopy. The linear concentration profile which is required for chemotactical movement is generated by diffusion and stable for at least 48 hours. #1.5 polymer coverslip, tissue culture treated, sterilized, 10 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-776 Supplier Ibidi USA Supplier No. 80326
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide Chemotaxis is a tool for investigation of chemotaxis and migration of nonadherent cells in 3D gel matrices and adherent cells on 2D surfaces. The chamber’s geometry is optimized for analyzing chemotaxis by video microscopy. The linear concentration profile which is required for chemotactical movement is generated by diffusion and stable for at least 48 hours. #1.5 polymer coverslip, tissue culture treated, sterilized, 10 pcs./box