Learn More
Ibidi USA U-SLIDE I 0.4 LUER GLASS BOTTM 1.5H (170 um +/- 5 um) D 263 M Schott glass, sterilized, 15 pcs./box
SDP
Catalog No. NC2226590
Encompass
The u-Slide I Luer is designed for cell culture under perfusion and all flow applications. Main applications are the simulation of blood vessels for arteriosclerosis research and applying defined shear stress and shear rates on cells inside the channel. The female Luers allow easy connections to tubing and pump systems. 450 ¿m high channel, #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC2226590 Supplier Ibidi USA Supplier No. 80177
The u-Slide I Luer is designed for cell culture under perfusion and all flow applications. Main applications are the simulation of blood vessels for arteriosclerosis research and applying defined shear stress and shear rates on cells inside the channel. The female Luers allow easy connections to tubing and pump systems. 450 ¿m high channel, #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilized, 15 pcs./box