Learn More
Ibidi USA µ-Slide I 0.4 Luer, Collagen IV - Collagen IV Coated Polymer Coverslip
SDP
Catalog No. 50305853
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide I Luer is designed for cell culture under perfusion and all flow applications. Main applications are the simulation of blood vessels for arteriosclerosis research and applying defined shear stress and shear rates on cells inside the channel. The female Luers allow easy connections to tubing and pump systems. 400 µm high channel, #1.5 polymer coverslip, collagen IV coated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-853 Supplier Ibidi USA Supplier No. 80172
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide I Luer is designed for cell culture under perfusion and all flow applications. Main applications are the simulation of blood vessels for arteriosclerosis research and applying defined shear stress and shear rates on cells inside the channel. The female Luers allow easy connections to tubing and pump systems. 400 µm high channel, #1.5 polymer coverslip, collagen IV coated, sterilized, 15 pcs./box