Learn More
Ibidi USA µ-Slide III 3 in 1, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305774
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide III 3in1 is designed for flow assays with different liquids merging into one channel. It can be connected to a pump and enables you to observe cells under switchable flow conditions. The design generates fluid-stable concentration profiles in the main channel for e.g. chemotaxis experiments. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-774 Supplier Ibidi USA Supplier No. 80316
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide III 3in1 is designed for flow assays with different liquids merging into one channel. It can be connected to a pump and enables you to observe cells under switchable flow conditions. The design generates fluid-stable concentration profiles in the main channel for e.g. chemotaxis experiments. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box