Learn More
Ibidi USA µ-Slide Microscopy Rack
SDP
Catalog No. 50305726
Encompass
µ-Slide Microscopy rack is a aluminum rack made to hold 4 µ-Slides for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Magnetic Lids sold seperately. aluminum, black, 127.5 x 85.5 x 12 mm
This item is not returnable. View return policy
Catalog No. 50-305-726 Supplier Ibidi USA Supplier No. 80030
µ-Slide Microscopy rack is a aluminum rack made to hold 4 µ-Slides for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Magnetic Lids sold seperately. aluminum, black, 127.5 x 85.5 x 12 mm