Learn More
Ibidi USA µ-Slide VI 0.1, ibiTreat - Tissue Culture Treated Coated Polymer Coverslip
SDP
Catalog No. 50305791
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide VI 0.1 is designed for flow assays in a minimal volume. It can be connected to a pump and enables you to observe cells under flow conditions. The small dimensions offer you the possibility to work with a minimum of cells. The convenient six channel format is ideal for the application of standard protocols for e.g. treatment, staining, and microscopy of living or fixed cells. 6 x 100 µm high channels, #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-791 Supplier Ibidi USA Supplier No. 80666
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide VI 0.1 is designed for flow assays in a minimal volume. It can be connected to a pump and enables you to observe cells under flow conditions. The small dimensions offer you the possibility to work with a minimum of cells. The convenient six channel format is ideal for the application of standard protocols for e.g. treatment, staining, and microscopy of living or fixed cells. 6 x 100 µm high channels, #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box