Learn More
Ibidi USA µ-Slide VI 0.4, Collagen IV - Collagen IV Coated Polymer Coverslip
SDP
Catalog No. 50305871
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The convenient six channel format of the µ-Slide VI 0.4 is ideal for static cell cultivation and the application of standard immunofluorescence protocols, like treatment, staining, and microscopy of living or fixed cells. Alternatively, the µ-Slide VI 0.4 can be connected to a pump and enables you to observe cells under flow conditions. 6 x 400 µm high channels, #1.5 polymer coverslip, collagen IV coated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-871 Supplier Ibidi USA Supplier No. 80602
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The convenient six channel format of the µ-Slide VI 0.4 is ideal for static cell cultivation and the application of standard immunofluorescence protocols, like treatment, staining, and microscopy of living or fixed cells. Alternatively, the µ-Slide VI 0.4 can be connected to a pump and enables you to observe cells under flow conditions. 6 x 400 µm high channels, #1.5 polymer coverslip, collagen IV coated, sterilized, 15 pcs./box