Learn More
Ibidi USA µ-Slide VI - Flat, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. 50305786
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide VI - flat is suited for quick cell tests a few hours after seeding. It is intended for optimization of experimental parameters like antibody dilution, seeding density or most effective drug concentration. Its flat dimensions allow convenient space-saving storage for your documentation in standard slide boxes. 6 x 400 µm high channels, #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-786 Supplier Ibidi USA Supplier No. 80621
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide VI - flat is suited for quick cell tests a few hours after seeding. It is intended for optimization of experimental parameters like antibody dilution, seeding density or most effective drug concentration. Its flat dimensions allow convenient space-saving storage for your documentation in standard slide boxes. 6 x 400 µm high channels, #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box