Learn More
Ibidi USA µ-Slide y-shaped, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305742
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide y-shaped can be easily connected to a pump via its Luer connectors. It enables you to grow cells under flow conditions with a bifurcation of 30 and 45 degree depending on the flow direction you choose. It is meant as a simulation system for blood vessels where the reaction of cells to a stimulus of your choice can be observed in real-time. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-742 Supplier Ibidi USA Supplier No. 80126
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide y-shaped can be easily connected to a pump via its Luer connectors. It enables you to grow cells under flow conditions with a bifurcation of 30 and 45 degree depending on the flow direction you choose. It is meant as a simulation system for blood vessels where the reaction of cells to a stimulus of your choice can be observed in real-time. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box