Learn More
Grainger Bessey Bar Clamp, Light Duty, 4 in. Opening, 330 lb. Force, 1 ea.
Catalog No. 502786456
Encompass_Preferred
This light-duty F-style bar clamp is designed for precise, small-scale applications, offering a 4 in. maximum jaw opening and 2 in. throat depth. Constructed with a zinc die cast frame, this tool provides a reliable 330 lb. clamping force, making it suitable for delicate projects requiring secure, non-marring pressure.
  • Features a clutch slide mechanism for efficient adjustments
  • Includes two plastic pressure caps to prevent surface damage
  • Equipped with swivel pads for clamping irregular or angled workpieces
  • Designed for compact size to reach confined spaces
  • Constructed with a wooden handle for comfortable operation
Catalog No. 50-278-6456 Supplier Grainger Supplier No. 19C740
This light-duty F-style bar clamp is designed for precise, small-scale applications, offering a 4 in. maximum jaw opening and 2 in. throat depth. Constructed with a zinc die cast frame, this tool provides a reliable 330 lb. clamping force, making it suitable for delicate projects requiring secure, non-marring pressure.
  • Features a clutch slide mechanism for efficient adjustments
  • Includes two plastic pressure caps to prevent surface damage
  • Equipped with swivel pads for clamping irregular or angled workpieces
  • Designed for compact size to reach confined spaces
  • Constructed with a wooden handle for comfortable operation