Learn More
Grainger Bessey U-Style Bar Clamp, Light Duty, 6 in. Capacity, 2 in. Throat Depth, 24/Pk.
Catalog No. 502787570
Encompass_Preferred
This light duty, U-style bar clamp is designed for woodworking applications. Constructed from durable zinc die cast material with a powder-coated finish, it provides a secure hold with a maximum clamping force of 330 lb and a 6 in. jaw opening.
  • Throat depth of 2 in.
  • Equipped with non-marring, replaceable swivel jaw pads
  • Features a wood handle and a clutch-based sliding mechanism
  • Includes removable plastic jaw caps for surface protection
  • Compact design with an approximate weight of 0.60 lb
Catalog No. 50-278-7570 Supplier Grainger Supplier No. 19C741
This light duty, U-style bar clamp is designed for woodworking applications. Constructed from durable zinc die cast material with a powder-coated finish, it provides a secure hold with a maximum clamping force of 330 lb and a 6 in. jaw opening.
  • Throat depth of 2 in.
  • Equipped with non-marring, replaceable swivel jaw pads
  • Features a wood handle and a clutch-based sliding mechanism
  • Includes removable plastic jaw caps for surface protection
  • Compact design with an approximate weight of 0.60 lb