Learn More
Drg International Inc DRG Ureaplasma Urealyticum IgA ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA5098R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Ureaplasma urealyticum in serum and plasma (EDTA or heparin plasma). This product is for Research Use Only. The DRG Ureaplasma urealyticum IgA ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyperimmune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with Ureaplasma urealyticum UreD protein. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7625