Learn More
Drg International Inc U. Urealyticum Igg Elisa
Supplier:  Drg International Inc EIA4623
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to ureaplasma urealyticum in serum and plasma. Ureaplasma urealyticum is a bacterium belonging to the family Mycoplasmataceae. This microorganism is part of the normal genital flora of both men and women. It is found in about 70% of sexually active humans. It can cause disease, including non-specific urethritis (NSU), infertility, chorioamnioitis, stillbirth, premature birth, and, in the perinatal period, pneumonia or meningitis. The DRG Ureaplasma urealyticum IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Ureaplasma urealyticum UreD protein. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Ureaplasma urealyticum-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
Catalog No. 50-194-7617