Learn More
Apexbio Technology LLC U0126 200mg
SDP
Catalog No. 5000089300
Encompass_Preferred
U0126 (CAS 109511-58-2) is a selective non-ATP competitive inhibitor of the MAPK/ERK kinase (MEK1/2) pathway interfering with downstream ERK1/2 activation By inhibiting MEK1/2 activity U0126 modulates cellular processes including proliferation differentiation and survival Additionally U0126 has been reported to suppress autophagy and mitophagy pathways This compound is widely used in biomedical research to investigate MAPK signaling dynamics as well as to explore the regulation of autophagic and mitochondrial quality control mechanisms in various cell models
Catalog No. 50-000-89300 Supplier Apexbio Technology LLC Supplier No. BA2003200
U0126 (CAS 109511-58-2) is a selective non-ATP competitive inhibitor of the MAPK/ERK kinase (MEK1/2) pathway interfering with downstream ERK1/2 activation By inhibiting MEK1/2 activity U0126 modulates cellular processes including proliferation differentiation and survival Additionally U0126 has been reported to suppress autophagy and mitophagy pathways This compound is widely used in biomedical research to investigate MAPK signaling dynamics as well as to explore the regulation of autophagic and mitochondrial quality control mechanisms in various cell models