Learn More
eMolecules​ U0126 | 109511-58-2 | MFCD01861186 | 25mg
Supplier:  eMolecules​ 562455925
Encompass_Preferred
Apollo Scientific | U0126 | 25mg | 562455925 | BIU1095 | 99.000 | 109511-58-2 | MFCD01861186 | 380.490 | C18H16N6S2
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-215-2011
 Restricted Item