Learn More
Apexbio Technology LLC U0126-EtOH, 25mg. Cas: 1173097-76-1 MFCD: MFCD11046040. MEK1/2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A133725
Encompass_Preferred
U0126-EtOH is a selective inhibitor of MEK1 and MEK2 with IC50 values of 70 nM and 60 nM, resepctively [1].U0126 was screened out as an anti-inflammatory agent that inhibited AP-1 transcription with IC50 value of 1M and had no interactions with GREs. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4692