Learn More
Medchemexpress LLC HY-12031 50mg , U0126-EtOH CAS:1173097-76-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1203150MG
Encompass_Preferred
HY-12031 50mg U0126-EtOH CAS: 1173097-76-1 U0126 (U0126-EtOH) is a potent, non-ATP competitive and selective MEK1 and MEK2 inhibitor, with IC50s of 72 nM and 58 nM, respectively. U0126 is an autophagy and mitophagy inhibitor[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9465