Learn More
Selleck Chemical LLC U0126-EtOH 100mg 1173097-76-1
SDP
Catalog No. 508422
Encompass_Preferred
U0126-EtOH is a highly selective inhibitor of MEK1/2 with IC50 of 0.07 ?M/0.06 ?M in cell-free assays, 100-fold higher affinity for ?N3-S218E/S222D MEK than PD98059. U0126 inhibits autophagy and mitophagy with antiviral activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-842-2 Supplier Selleck Chemical LLC Supplier No. S1102100MG
U0126-EtOH is a highly selective inhibitor of MEK1/2 with IC50 of 0.07 ?M/0.06 ?M in cell-free assays, 100-fold higher affinity for ?N3-S218E/S222D MEK than PD98059. U0126 inhibits autophagy and mitophagy with antiviral activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.