Learn More
Montana Molecular Green Upward cADDis cAMP sensor Big Sky BacMam 5mL
SDP
Catalog No. 502815256
Encompass_Preferred
Detect cAMP signaling kinetics in living cells with the Green Upward cADDis fluorescent cAMP assay. Increase in fluorescence indicates increase in cAMP. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an updated version of U0200G, with improved expression efficiency from Big Sky BacMam.
Catalog No. 50-281-5256 Supplier Montana Molecular Supplier No. U0250G005
Detect cAMP signaling kinetics in living cells with the Green Upward cADDis fluorescent cAMP assay. Increase in fluorescence indicates increase in cAMP. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an updated version of U0200G, with improved expression efficiency from Big Sky BacMam.