Learn More
Montana Molecular Green Upward DAG sensor Big Sky BacMam 5mL
SDP
Catalog No. 502815257
Encompass_Preferred
Detect DAG and Gq signaling kinetics in living cells with the Green Upward fluorescent DAG assay. Increase in fluorescence indicates increase in DAG. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an updated version of product U0300G, with improved expression efficiency from Big Sky BacMam.
Catalog No. 50-281-5257 Supplier Montana Molecular Supplier No. U0350G005
Detect DAG and Gq signaling kinetics in living cells with the Green Upward fluorescent DAG assay. Increase in fluorescence indicates increase in DAG. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an updated version of product U0300G, with improved expression efficiency from Big Sky BacMam.