Learn More
ATCC U-138 MG; Glioblastoma; Human (Homo sapiens)
Catalog No. 502382472
Encompass
U-138 MG is a polygonal cell that was isolated from the brain of a 47-year-old, White, male with glioblastoma, grade IV This is one of a number of cell lines derived from malignant gliomas (see also ATCC HTB-14 and ATCC HTB-15 ) by J. Ponten and associates from 1966 to 1969. The cell line U-138 MG can be used in neuroscience research.
Catalog No. 50-238-2472 Supplier ATCC Supplier No. HTB16
U-138 MG is a polygonal cell that was isolated from the brain of a 47-year-old, White, male with glioblastoma, grade IV This is one of a number of cell lines derived from malignant gliomas (see also ATCC HTB-14 and ATCC HTB-15 ) by J. Ponten and associates from 1966 to 1969. The cell line U-138 MG can be used in neuroscience research.