Learn More
TARGETMOL CHEMICALS INC U18666A 25MG
SDP
Catalog No. 502581116
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. U18666A is a cell-permeable drug and is a cholesterol synthesis and transport inhibitor. It inhibits replication of Ebola virus dengue virus and human hepatitis C virus. purity: 99%
Catalog No. 50-258-1116 Supplier TARGETMOL CHEMICALS INC Supplier No. T1719025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. U18666A is a cell-permeable drug and is a cholesterol synthesis and transport inhibitor. It inhibits replication of Ebola virus dengue virus and human hepatitis C virus. purity: 99%