Learn More
Selleck Chemical LLC U18666A-5mg
SDP
Catalog No. 502389616
Encompass_Preferred
U18666A (U18) is an inhibitor of intra-cellular cholesterol transport that inhibits dengue virus entry and replication.
Catalog No. 50-238-9616 Supplier Selleck Chemical LLC Supplier No. S96695MG
U18666A (U18) is an inhibitor of intra-cellular cholesterol transport that inhibits dengue virus entry and replication.