Learn More
Aviva Systems Biology Corp U2AF1 Antibody - N-terminal region (ARP40326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against U2AF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human U2AF1. Alternate names: DKFZp313J1712, FP793, RN, RNU2AF1, U2AF35, U2AFBPApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57393