Learn More
Aviva Systems Biology Corp U2AF2 Antibody - C-terminal region (ARP40285_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40285_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against U2AF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human U2AF2. Alternate names: U2AF65ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-284-863