Learn More
Eurofins Discoverx PathHunter U2OS ACVR1/ACVR2 Dimerization Cell Line, 2 vials. (93-0906C3)
Catalog No. 502001540
Encompass
The PathHunter ACVR1/ACVR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-7) induced dimerization of ACVR1/ACVR2 receptors. This product uses EFC technology. The cell line overexpresses truncated ACVR1 target protein fused to PK alongside truncated ACVR2 target protein fused to EA. Upon ligand-induced dimerization of ACVR1/ACVR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1540 Supplier Eurofins Discoverx Supplier No. 930906C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter ACVR1/ACVR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-7) induced dimerization of ACVR1/ACVR2 receptors. This product uses EFC technology. The cell line overexpresses truncated ACVR1 target protein fused to PK alongside truncated ACVR2 target protein fused to EA. Upon ligand-induced dimerization of ACVR1/ACVR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.