Learn More
Eurofins Discoverx PathHunter U2OS ACVR1B/BMPR2 Dimerization Cell Line, 2 vials. (93-1072C3)
Catalog No. 502001981
Encompass
The PathHunter ACVR1B/BMPR2  Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. activin A) induced dimerization of ACVR1B/BMPR2  receptors. This product uses EFC technology. The cell line included overexpresses ACVR1B target protein fused to PK alongside BMPR2  target protein fused to EA. Upon ligand-induced dimerization of ACVR1B/BMPR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1981 Supplier Eurofins Discoverx Supplier No. 931072C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter ACVR1B/BMPR2  Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. activin A) induced dimerization of ACVR1B/BMPR2  receptors. This product uses EFC technology. The cell line included overexpresses ACVR1B target protein fused to PK alongside BMPR2  target protein fused to EA. Upon ligand-induced dimerization of ACVR1B/BMPR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.