Learn More
Eurofins Discoverx PathHunter U2OS ACVR1C/ACVR2B Dimerization Cell Line, 2 vials. (93-1047C3)
Supplier:  Eurofins Discoverx 931047C3
Encompass
The PathHunter ACVR1C/ACVR2B Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. activin B) induced dimerization of ACVR1C/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVR2B target protein fused to PK alongside ACVR1C target protein fused to EA. Upon ligand-induced dimerization of ACVR1C/ACVR2B, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1930
Add to Cart
Request a Quote