Learn More
Eurofins Discoverx PathHunter U2OS ACVRL1/BMPR2 Dimerization Cell Line, 2 vials. (93-0962C3)
Catalog No. 502001680
Encompass
The PathHunter ACVRL1/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. GDF2/BMP9) induced dimerization of ACVRL1/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses ACVRL1 target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of ACVRL1/BMPR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1680 Supplier Eurofins Discoverx Supplier No. 930962C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter ACVRL1/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. GDF2/BMP9) induced dimerization of ACVRL1/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses ACVRL1 target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of ACVRL1/BMPR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.