Learn More
Eurofins Discoverx PathHunter U2OS AVPR1A β-Arrestin Cell Line, 2 vials. (93-0587C3)
Supplier:  Eurofins Discoverx 930587C3
Encompass
The PathHunter AVPR1A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. vasopressin) based activation of AVPR1A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged AVPR1A and EA-tagged β-Arrestin-2. Activation of AVPR1A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0891
Add to Cart
Request a Quote