Learn More
Eurofins Discoverx PathHunter U2OS AVPR2(S167T) ENDO-EA Pharmacotrafficking Assay, 2 vials. (93-0992C3)
Catalog No. 502001764
Encompass
The PathHunter U2OS AVPR2(S167T) ENDO-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting AVPR2. This cell line enables assessment of chaperone-based stabilization of AVPR2(S167T) via detection of rescued membrane-endosome trafficking. The included cell line overexpresses ProLink-tagged mutant AVPR2(S167T), which is misfolded and retained in the ER. The cell line also expresses endosome-localized EA. When a pharmacochaperone stabilizes the mutant AVPR2, it is trafficked to the membrane and then internalized to the endosome, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1764 Supplier Eurofins Discoverx Supplier No. 930992C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS AVPR2(S167T) ENDO-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting AVPR2. This cell line enables assessment of chaperone-based stabilization of AVPR2(S167T) via detection of rescued membrane-endosome trafficking. The included cell line overexpresses ProLink-tagged mutant AVPR2(S167T), which is misfolded and retained in the ER. The cell line also expresses endosome-localized EA. When a pharmacochaperone stabilizes the mutant AVPR2, it is trafficked to the membrane and then internalized to the endosome, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.