Learn More
Eurofins Discoverx InCELL Hunter U2OS Bcl2-Bax Protein Binding Cell Line, 2 vials. (96-0010C3)
Catalog No. 502004200
Encompass
The InCELL Hunter U2OS Bcl2-Bax Protein Binding Cell Line provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2 target in an untagged form and Bax binding partner fused to ProLabel. When a compound engages the Bcl2 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-4200 Supplier Eurofins Discoverx Supplier No. 960010C3
Add to Cart
Add to Cart
Request a Quote
The InCELL Hunter U2OS Bcl2-Bax Protein Binding Cell Line provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2 target in an untagged form and Bax binding partner fused to ProLabel. When a compound engages the Bcl2 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.