Learn More
Eurofins Discoverx PathHunter U2OS BLK Activity Assay, 2 vials. (93-0882C3)
Catalog No. 502001482
Encompass
The PathHunter U2OS BLK Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the BLK kinase domain. The cell line is engineered to express the catalytic domain from BLK fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the GHR-PK. Activation of the GHR-PK induces receptor phosphorylation by the BLK catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1482 Supplier Eurofins Discoverx Supplier No. 930882C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS BLK Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the BLK kinase domain. The cell line is engineered to express the catalytic domain from BLK fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the GHR-PK. Activation of the GHR-PK induces receptor phosphorylation by the BLK catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.