Learn More
Eurofins Discoverx PathHunter U2OS BMPR1A/ACVR2A Dimerization Cell Line, 2 vials. (93-1037C3)
Supplier:  Eurofins Discoverx 931037C3
Encompass
The PathHunter BMPR1A/ACVR2A Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-4) induced dimerization of BMPR1A/ACVR2A receptors. This product uses EFC technology. The cell line included overexpresses BMPR1A target protein fused to PK alongside ACVR2A target protein fused to EA. Upon ligand-induced dimerization of BMPR1A/ACVR2A, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1901
Only null left
Add to Cart
Request a Quote