Learn More
Eurofins Discoverx PathHunter U2OS BMPR1B/BMPR2 Dimerization Cell Line, 2 vials. (93-1053C3)
Catalog No. 502001944
Encompass
The PathHunter BMPR1B/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-6) induced dimerization of BMPR1B/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR1B target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of BMPR1B/BMPR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1944 Supplier Eurofins Discoverx Supplier No. 931053C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter BMPR1B/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-6) induced dimerization of BMPR1B/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR1B target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of BMPR1B/BMPR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.