Learn More
Eurofins Discoverx PathHunter U2OS c-KIT Functional Assay, 2 vials. (93-0853C3)
Supplier:  Eurofins Discoverx 930853C3
Encompass
The PathHunter U2OS c-KIT Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated c-KIT dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged c-KIT and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the cynomolgus version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1426
Add to Cart
Request a Quote