Learn More
Eurofins Discoverx U2OS CASR Gq Cell Line, 2 vials. (95-1014C3)
Catalog No. 502004076
Encompass
The CASR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. extracellular calcium) based activation of CASR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CASR, a naturally Gq coupled GPCR. Upon activation of CASR, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4076 Supplier Eurofins Discoverx Supplier No. 951014C3
Add to Cart
Add to Cart
Request a Quote
The CASR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. extracellular calcium) based activation of CASR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CASR, a naturally Gq coupled GPCR. Upon activation of CASR, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.