Learn More
Eurofins Discoverx PathHunter U2OS CHRM1 β-Arrestin Cell Line, 2 vials. (93-0859C3)
Supplier:  Eurofins Discoverx 930859C3
Encompass
The PathHunter CHRM1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. acetylcholine) based activation of CHRM1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CHRM1 and EA-tagged β-Arrestin-2. Activation of CHRM1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1435
Add to Cart
Request a Quote